missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-(trifluoromethyl)benzoic acid, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | 4-Bromo-2-(trifluoromethyl)benzoic acid |
| CAS | 320-31-0 |
| Molecular Formula | C8H4BrF3O2 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C8H4BrF3O2/c9-4-1-2-5(7(13)14)6(3-4)8(10,11)12/h1-3H,(H,13,14) |
| SMILES | OC(=O)c1ccc(Br)cc1C(F)(F)F |
| IUPAC Name | 4-bromo-2-(trifluoromethyl)benzoic acid |
| Molecular Weight (g/mol) | 269.02 |
| Formula Weight | 267.9347 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction