missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,3,4,4-tetrafluoro-1-butene, 98%, Thermo Scientific™

Product Code. 11375015
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11375015 5 g 5g
11365015 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11375015 Supplier Thermo Scientific Alfa Aesar Supplier No. L18492.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 18599-22-9
Molecular Formula C4H3BrF4
Molecular Weight (g/mol) 206.97
MDL Number MFCD00042189
InChI Key GVCWGFZDSIWLMO-UHFFFAOYSA-N
Synonym 4-bromo-3,3,4,4-tetrafluoro-1-butene, 1-butene, 4-bromo-3,3,4,4-tetrafluoro, unii-t08c3oeq6e, t08c3oeq6e, 4-bromo-3,3,4,4-tetrafluorobutene-1, 1-butene,4-bromo-3,3,4,4-tetrafluoro, acmc-209enf, gvcwgfzdsiwlmo-uhfffaoysa, 4-bromo-3,3,4,4-tetrafluorobutene, 1-bromo-1,1,2,2-tetrafluoro-3-butene
PubChem CID 87721
IUPAC Name 4-bromo-3,3,4,4-tetrafluorobut-1-ene
SMILES FC(F)(Br)C(F)(F)C=C

Specifica

Density 1.627
Boiling Point 55°C to 57°C
Flash Point None
Linear Formula BrCF2CF2CH=CH2
Refractive Index 1.354
Quantity 5 g
Beilstein 1856181
Formula Weight 206.97
Percent Purity 98%
Chemical Name or Material 4-Bromo-3,3,4,4-tetrafluoro-1-butene
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.