missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dimethyl-1H-pyrazole, 98%

Product Code. 11489313
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489313 100 g 100g
11469313 5 g 5g
11479313 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489313 Supplier Thermo Scientific Alfa Aesar Supplier No. A11759.22
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3398-16-1 | C5H7BrN2 | 175.029 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3398-16-1
Molecular Formula C5H7BrN2
Molecular Weight (g/mol) 175.029
MDL Number MFCD00005242
InChI Key RISOHYOEPYWKOB-UHFFFAOYSA-N
Synonym 4-bromo-3,5-dimethylpyrazole, pyrazole, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromopyrazole, 1h-pyrazole, 4-bromo-3,5-dimethyl, 3.5-dimethyl-4-bromopyrazole, acmc-1cnce, 5-23-05-00154 beilstein handbook reference, ksc495g9b, 4-bromo-3,5-dimethyl pyrazole, 4-bromo-3,5-dimethyl-pyrazole
PubChem CID 76937
IUPAC Name 4-bromo-3,5-dimethyl-1H-pyrazole
SMILES CC1=C(C(=NN1)C)Br

Specifications

Melting Point 118°C to 123°C
Quantity 100 g
Beilstein 108534
Formula Weight 175.03
Percent Purity 99%
Chemical Name or Material 4-Bromo-3,5-dimethyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.