missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dimethylpyrazole, 99%, ACROS Organics™

Product Code. 10353722 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10353722 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10353722 Supplier Thermo Scientific Acros Supplier No. 354290100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 3398-16-1
CAS Min % 98.5
CAS Max % 100.0
Melting Point 120°C to 124°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C5H7BrN2
MDL Number MFCD00005242
Quantity 10 g
Synonym 4-bromo-3,5-dimethylpyrazole, pyrazole, 4-bromo-3,5-dimethyl, 3,5-dimethyl-4-bromopyrazole, 1h-pyrazole, 4-bromo-3,5-dimethyl, 3.5-dimethyl-4-bromopyrazole, acmc-1cnce, 5-23-05-00154 beilstein handbook reference, ksc495g9b, 4-bromo-3,5-dimethyl pyrazole, 4-bromo-3,5-dimethyl-pyrazole
InChI Key RISOHYOEPYWKOB-UHFFFAOYSA-N
SMILES CC1=C(C(=NN1)C)Br
IUPAC Name 4-bromo-3,5-dimethyl-1H-pyrazole
Molecular Weight (g/mol) 175.029
PubChem CID 76937
Formula Weight 175.03
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3, 5-dimethylpyrazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.