missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-phenyl-1H-pyrazol-5-amine, ≥95%, Thermo Scientific™

Product Code. 10123864
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10123864 1 g Amber Glass Bottle 1g
10377064 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10123864 Supplier Thermo Scientific Maybridge Supplier No. KM00411DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 2845-78-5
Molecular Formula C9H8BrN3
Molecular Weight (g/mol) 238.09
MDL Number MFCD00082666
InChI Key QTNVXMOPTHGCII-UHFFFAOYSA-N
Synonym 4-bromo-3-phenyl-1h-pyrazol-5-amine, 3-amino-4-bromo-5-phenylpyrazole, 4-bromo-5-phenyl-2h-pyrazol-3-amine, 3-amino-4-bromo-5-phenyl-1h-pyrazole, 1h-pyrazol-3-amine,4-bromo-5-phenyl, acmc-20amn3, maybridge1_004579, 4-bromo-3-phenylpyrazole-5-ylamine, 4-bromo-5-phenylpyrazole-3-ylamine, 4-bromo-3-phenyl-1h-pyrazole-5-amine
PubChem CID 594319
IUPAC Name 4-bromo-5-phenyl-1H-pyrazol-3-amine
SMILES NC1=NNC(=C1Br)C1=CC=CC=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 238.09
Percent Purity 95%
Chemical Name or Material 4-Bromo-3-phenyl-1H-pyrazol-5-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.