missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-(trifluoromethyl)-1H-pyrazole, 97%, Thermo Scientific™

Product Code. 10735842 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10735842 5 g 5g
10037619 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10735842 Supplier Thermo Scientific Acros Supplier No. 438040050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 19968-17-3
Molecular Formula C4H2BrF3N2
Molecular Weight (g/mol) 214.97
InChI Key JTHNMRUVJDWVMJ-UHFFFAOYSA-N
Synonym 4-bromo-3-trifluoromethyl-1h-pyrazole, 4-bromo-5-trifluoromethyl-1h-pyrazole, 1h-pyrazole, 4-bromo-3-trifluoromethyl, 1h-pyrazole, 4-bromo-5-trifluoromethyl, 1h-pyrazole,4-bromo-3-trifluoromethyl, 4-bromo-3-trifluoromethyl pyrazole, 4-bromo-3-trifluoromethyl-2h-pyrazole, acmc-209f4m, 4-bromo-3 trifluoromethyl-1h-pyrazole
PubChem CID 7147398
IUPAC Name 4-bromo-5-(trifluoromethyl)-1H-pyrazole
SMILES C1=NNC(=C1Br)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 112°C to 114°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 214.97
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-(trifluoromethyl)-1H-pyrazole
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-3-(trifluoromethyl)-1H-pyrazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.