missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-5-chlorothiophene-2-sulfonyl chloride, ≥95%, Thermo Scientific™

Product Code. 10640923
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10640923 250 mg 250mg
10283082 1 g 1g
10307404 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10640923 Supplier Thermo Scientific Maybridge Supplier No. SEW02652CB
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 166964-35-8
Molecular Formula C4HBrCl2O2S2
Molecular Weight (g/mol) 295.974
MDL Number MFCD00052094
InChI Key PABOKJJDABSQCK-UHFFFAOYSA-N
Synonym 3-bromo-2-chlorothiophene-5-sulfonyl chloride, 4-bromo-5-chlorothiophene-2-sulphonyl chloride, 4-bromo-5-chloro-2-thiophenesulfonyl chloride, 4-bromo-5-chlorothiophene-2-sulphonylchloride, 3-bromo-2-chlorothiophene-5-sulphonyl chloride, 2-thiophenesulfonylchloride, 4-bromo-5-chloro, pubchem5475, 4-bromo-5-chloro-thiophene-2-sulfonyl chloride, 4-bromo-5-chlorothiophene-2-sulfonyl, 4-bromanyl-5-chloranyl-thiophene-2-sulfonyl chloride
PubChem CID 2797220
IUPAC Name 4-bromo-5-chlorothiophene-2-sulfonyl chloride
SMILES C1=C(SC(=C1Br)Cl)S(=O)(=O)Cl

Specifica

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 295.99
Percent Purity 95%
Chemical Name or Material 4-Bromo-5-chlorothiophene-2-sulfonyl chloride
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.