missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N-(2,4-dimethylphenyl)benzamide, 97%, Thermo Scientific™

Product Code. 15493788
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493788 250 mg 250mg
15483788 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493788 Supplier Thermo Scientific Alfa Aesar Supplier No. H57284.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 282091-66-1
Molecular Formula C15H14BrNO
Molecular Weight (g/mol) 304.19
MDL Number MFCD00436496
InChI Key CGUNDMXTDHRITR-UHFFFAOYSA-N
Synonym 4-bromo-n-2,4-dimethylphenyl benzamide, 4-bromo-n-2,4-dimethyl-phenyl-benzamide
PubChem CID 774375
IUPAC Name 4-bromo-N-(2,4-dimethylphenyl)benzamide
SMILES CC1=CC(C)=C(NC(=O)C2=CC=C(Br)C=C2)C=C1

Spezifikation

Quantity 250 mg
Formula Weight 304.19
Percent Purity 97%
Chemical Name or Material 4-Bromo-N-(2,4-dimethylphenyl)benzamide
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.