missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N-methylaniline, 97%

Product Code. 15401478
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15401478 1 g 1g
15411478 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15401478 Supplier Thermo Scientific Alfa Aesar Supplier No. H55273.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6911-87-1 | C7H8BrN | 186.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6911-87-1
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD03030391
InChI Key AYVPVDWQZAAZCM-UHFFFAOYSA-N
PubChem CID 2757052
IUPAC Name 4-bromo-N-methylaniline
SMILES CNC1=CC=C(Br)C=C1

Specifications

Density 1.482 g/mL
Boiling Point 259°C to 260°C
Refractive Index 1.613
Quantity 1 g
Synonym 4-bromo-n-methylbenzenamine, benzenamine, 4-bromo-n-methyl, n-methyl-4-bromoaniline, benzenamine,4-bromo-n-methyl, 4bromo-n-methylaniline, 4-bromo-n-methyl-aniline, n-methyl-4-bromo-aniline, acmc-1cfc7, 4-bromanyl-n-methyl-aniline, 4-bromo-phenyl-methyl-amine
Formula Weight 186.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Bromo-N-methylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.