missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromoacetophenone, 98%

Product Code. 11318561 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
Product Code. Quantity Packaging unitSize
11318561 500 g Glass bottle 500g
10509810 25 g Glass bottle 25g
10605142 100 g Glass bottle 100g
3 options
This item is not returnable. View return policy

Product Code. 11318561

Brand: Thermo Scientific Acros 106595000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-90-1 | C8H7BrO | 199.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-90-1
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD00000105
InChI Key WYECURVXVYPVAT-UHFFFAOYSA-N
Synonym 4'-bromoacetophenone, 1-4-bromophenyl ethanone, p-bromoacetophenone, 4-bromoacetophenone, ethanone, 1-4-bromophenyl, 1-4-bromophenyl ethan-1-one, 1-acetyl-4-bromobenzene, methyl p-bromophenyl ketone, p-bromophenyl methyl ketone, acetophenone, 4'-bromo
PubChem CID 7466
IUPAC Name 1-(4-bromophenyl)ethanone
SMILES CC(=O)C1=CC=C(Br)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 48°C to 52°C
Color White to Yellow
Boiling Point 255°C
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4COCH3
Quantity 500 g
Beilstein 07,283
Merck Index 15,141
Solubility Information Solubility in water: insoluble
Formula Weight 199.05
Percent Purity 98%
Physical Form Crystalline Flakes or Crystals
Chemical Name or Material 4'-Bromoacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.