missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromoacetophenone, 98%

Product Code. 11444903
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444903 25 g 25g
11454903 100 g 100g
11464903 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444903 Supplier Thermo Scientific Alfa Aesar Supplier No. A12222.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-90-1 | C8H7BrO | 199.05 g/mol

Labeled 4’-Bromoacetophenone possess activity against positive phototaxis of Chlamydomonas cells. A fundamental starting material for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Labeled 4’-Bromoacetophenone possess activity against positive phototaxis of Chlamydomonas cells. A fundamental starting material for organic synthesis.

Solubility
Soluble in Chloroform. Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-90-1
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD00000105
InChI Key WYECURVXVYPVAT-UHFFFAOYSA-N
Synonym 4'-bromoacetophenone, 1-4-bromophenyl ethanone, p-bromoacetophenone, 4-bromoacetophenone, ethanone, 1-4-bromophenyl, 1-4-bromophenyl ethan-1-one, 1-acetyl-4-bromobenzene, methyl p-bromophenyl ketone, p-bromophenyl methyl ketone, acetophenone, 4'-bromo
PubChem CID 7466
IUPAC Name 1-(4-bromophenyl)ethanone
SMILES CC(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 49°C to 53°C
Boiling Point 255°C to 256°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 386015
Merck Index 14,1403
Solubility Information Soluble in Chloroform. Insoluble in water.
Formula Weight 199.05
Percent Purity 98%
Chemical Name or Material 4'-Bromoacetophenone
compliance-icons
Product Identifier
  • 4'-Bromoacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4'-Bromoacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.