missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromoacetophenone, 98%

Product Code. 11454903
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454903 100 g 100g
11444903 25 g 25g
11464903 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. Consulta la politica di reso
Codice prodotto. 11454903 Fornitore Thermo Scientific Alfa Aesar N. del fornitore A12222.22
 Restricted Item
In Stock
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 99-90-1 | C8H7BrO | 199.05 g/mol

Labeled 4’-Bromoacetophenone possess activity against positive phototaxis of Chlamydomonas cells. A fundamental starting material for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Labeled 4’-Bromoacetophenone possess activity against positive phototaxis of Chlamydomonas cells. A fundamental starting material for organic synthesis.

Solubility
Soluble in Chloroform. Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent, bases.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 99-90-1
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD00000105
InChI Key WYECURVXVYPVAT-UHFFFAOYSA-N
Synonym 4'-bromoacetophenone, 1-4-bromophenyl ethanone, p-bromoacetophenone, 4-bromoacetophenone, ethanone, 1-4-bromophenyl, 1-4-bromophenyl ethan-1-one, 1-acetyl-4-bromobenzene, methyl p-bromophenyl ketone, p-bromophenyl methyl ketone, acetophenone, 4'-bromo
PubChem CID 7466
IUPAC Name 1-(4-bromophenyl)ethanone
SMILES CC(=O)C1=CC=C(Br)C=C1

Specifica

Melting Point 49°C to 53°C
Boiling Point 255°C to 256°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 386015
Merck Index 14,1403
Solubility Information Soluble in Chloroform. Insoluble in water.
Formula Weight 199.05
Percent Purity 98%
Chemical Name or Material 4'-Bromoacetophenone

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.