missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzenesulfonyl Chloride, TRC

Product Code. 30337606
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30337606 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337606 Supplier Toronto Research Chemicals Supplier No. TRCB68017025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material 4-Bromobenzenesulfonyl Chloride
CAS 98-58-8
Molecular Formula C6H4BrClO2S
Quantity 25 g
Synonym p-Bromobenzenesulfonyl Chloride, (4-Bromophenyl)chlorosulfone, 4-Bromobenzene-1-sulfonyl Chloride, 4-Bromophenylsulfonyl Chloride, 4'-Bromobenzenesulfonyl Chloride, Brosyl Chloride, NSC 4506, p-Bromophenylsulfonyl Chloride, p-Brosyl Chloride
InChI Formula InChI=1S/C6H4BrClO2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H
SMILES ClS(=O)(=O)c1ccc(Br)cc1
IUPAC Name 4-bromobenzenesulfonyl chloride
Molecular Weight (g/mol) 255.52
Formula Weight 253.8804
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.