missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzoic Acid, TRC

Product Code. 30331390
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30331390 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331390 Supplier Toronto Research Chemicals Supplier No. TRCB68030025G
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Bromobenzoic Acid
CAS 586-76-5
Molecular Formula C7H5BrO2
Quantity 25 g
Synonym 4-Bromobenzoic Acid, p-Bromobenzoic Acid, 4-Bromobenzenecarboxylic Acid, 4-Bromobenzoic Acid, NSC 143357, NSC 17582, NSC 176126, p-Bromobenzenecarboxylic Acid, p-Bromobenzoic cid, p-Carboxybromobenzene
InChI Formula InChI=1S/C7H5BrO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10)
SMILES OC(=O)c1ccc(Br)cc1
IUPAC Name 4-bromobenzoic acid
Molecular Weight (g/mol) 201.02
Formula Weight 199.9473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.