missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobenzyl-2,3,5,6-d4 Bromide, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 4-Bromobenzyl Bromide-2,3,5,6-D4 |
| CAS | 2708280-84-4 |
| Molecular Formula | C7 D4 H2 Br2 |
| Quantity | 0.5 g |
| Synonym | 3-Bromo-6-bromomethylbenzene-1,2,4,5-d4, 4-Bromobenzyl-d4 Bromide, 4-Bromobenzyl-2,3,5,6-d4 Bromide, 1-Bromo-4-(bromomethyl)-2,3,5,6-tetradeuteriobenzene, 1-Bromomethyl-4-bromobenzene-2,3,5,6-d4, 4-Bromobenzyl Bromide-2,3,5,6-D4 |
| InChI Formula | InChI=1 S/C7H6Br2/c8-5-6-1-3-7(9)4-2-6/h1-4 H,5H2/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(CBr)c([2 H])c([2 H])c1Br |
| IUPAC Name | 1-bromo-4-(bromomethyl)-2,3,5,6-tetradeuteriobenzene |
| Molecular Weight (g/mol) | 253.9552 |
| Formula Weight | 251.9087 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction