missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzyl Bromide, TRC

Product Code. 30333364
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30333364 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333364 Supplier Toronto Research Chemicals Supplier No. TRCB679955100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Bromobenzyl Bromide
CAS 589-15-1
Molecular Formula C22 H28 O4
Quantity 100 g
Synonym p,alpha-Dibromo-toluene, 1-(Bromomethyl)-4-bromobenzene, 1-Bromo-4-(bromomethyl)benzene, 4-(Bromomethyl)bromobenzene, 4-Bromo-1-bromomethylbenzene, 4-Bromobenzyl bromide, NSC 113584, p-Bromobenzyl bromide, alpha,p-Dibromotoluene
InChI Formula InChI=1S/C22H28O4/c1-4-22(26-13(2)23)10-8-19-17-12-20(25)18-11-14(24)5-6-15(18)16(17)7-9-21(19,22)3/h1,11,15-17,19-20,25H,5-10,12H2,2-3H3/t15-,16+,17-,19-,20-,21+,22?/m1/s1
SMILES CC(=O)OC1(CC[C@@H]2[C@@H]3C[C@@H](O)C4=CC(=O)CC[C@@H]4[C@@H]3CC[C@]12C)C#C
IUPAC Name [(6R,8R,9R,10R,13S,14R)-17-ethynyl-6-hydroxy-13-methyl-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 356.455
Formula Weight 356.199
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.