missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzylamine, 97+%

Product Code. 10522544 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10522544 25 g Glass bottle 25g
10500912 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10522544 Supplier Thermo Scientific Acros Supplier No. 350530250
 Restricted Item
Estimated Shipment: 30-10-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format
Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 3959-07-7 | C7H8BrN | 186.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 3959-07-7
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD00047931
InChI Key XRNVSPDQTPVECU-UHFFFAOYSA-N
SMILES NCC1=CC=C(Br)C=C1

Especificaciones

CAS Min % 98.5
CAS Max % 100.0
Melting Point 25.0°C
Color Colorless
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CH2NH2
Refractive Index 1.5855 to 1.5875
Quantity 25 g
Beilstein 12, III, 2350
Synonym quinoline, 4-bromo, 4-bromo-quinoline, pubchem7115, acmc-1aijg, 4-bromoquinoline, ksc222e1b
IUPAC Name 1-(4-bromophenyl)methanamine
PubChem CID 11735918
Formula Weight 186.05
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material 4-Bromobenzylamine
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 4-Bromobenzylamine
Palabra de advertencia
  • Danger
Clasificación
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Indicaciones de peligro
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.