missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutyric Acid, TRC

Product Code. 30346687
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 30346687

Brand: Toronto Research Chemicals TRCB68204510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Bromobutyric Acid
CAS 2623-87-2
Molecular Formula C4H7BrO2
Quantity 10 g
Synonym 4-Bromobutanoic Acid, 4-Bromobutyric Acid, Carboxypropyl Bromide, NSC 99322, γ-Bromobutyric Acid
InChI Formula InChI=1S/C4H7BrO2/c5-3-1-2-4(6)7/h1-3H2,(H,6,7)
SMILES OC(=O)CCCBr
IUPAC Name 4-bromobutanoic acid
Molecular Weight (g/mol) 167
Formula Weight 165.9629
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.