missing translation for 'onlineSavingsMsg'
Learn More

4-Bromochlorobenzene, 99%

Product Code. 10148003 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
100 g
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
500g
50g
This item is not returnable. View return policy

Product Code. 10148003

Brand: Thermo Scientific Acros 106831000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-39-8 | C6H4BrCl | 191.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-39-8
Molecular Formula C6H4BrCl
Molecular Weight (g/mol) 191.45
MDL Number MFCD00000600
InChI Key NHDODQWIKUYWMW-UHFFFAOYSA-N
Synonym 4-bromochlorobenzene, p-bromochlorobenzene, 4-chlorobromobenzene, p-chlorobromobenzene, p-chlorophenyl bromide, benzene, 1-bromo-4-chloro, 4-chlorophenyl bromide, 1-chloro-4-bromobenzene, 4-chloro-1-bromobenzene, p-bromoclorobenzene
PubChem CID 7806
IUPAC Name 1-bromo-4-chlorobenzene
SMILES ClC1=CC=C(Br)C=C1

Specifications

Melting Point 64.0°C to 67.0°C
CAS Min % 98.5
Color White
CAS Max % 100.0
Density 1.6510g/mL
Boiling Point 196.0°C
Quantity 100 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder
Assay Percent Range 98.5% min. (GC)
Specific Gravity 1.651
Linear Formula BrC6H4Cl
Beilstein 05, 209
Solubility Information Solubility in water: 0.045g/L (25°C). Other solubilities: soluble in most organic solvents
Formula Weight 191.45
Percent Purity 99%
Chemical Name or Material 4-Bromochlorobenzene, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.