missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoindole, 98%

Product Code. 11388716
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388716 5 g 5g
11398716 250 mg 250mg
11378716 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388716 Supplier Thermo Scientific Alfa Aesar Supplier No. L19417.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 52488-36-5 | C8H6BrN | 196.047 g/mol

4-Bromoindole is a potential inhibitor of GSK-3. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromoindole is a potential inhibitor of GSK-3. It is also used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Air & Light Sensitive. Store at 4°C. Store in the dark. Store under dry inert gas. Protect from heat. Incompatible with heat. light, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52488-36-5
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD00671502
InChI Key GRJZJFUBQYULKL-UHFFFAOYSA-N
Synonym 4-bromoindole, 1h-indole, 4-bromo, 4-bromo-indole, pubchem7449, acmc-1amo5, ksc269g6j, grjzjfubqyulkl-uhfffaoysa
PubChem CID 676494
IUPAC Name 4-bromo-1H-indole
SMILES C1=CC2=C(C=CN2)C(=C1)Br

Specifications

Melting Point 17°C
Density 1.563
Boiling Point 88°C to 90°C (0.1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6545
Quantity 5 g
Beilstein 114891
Sensitivity Air and light sensitive
Solubility Information Not miscible in water.
Formula Weight 196.05
Percent Purity 98%
Chemical Name or Material 4-Bromoindole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.