missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoisoindoline hydrochloride, 97%

Product Code. 10575870 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10575870 1 g Glass bottle 1g
10725652 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10575870 Supplier Thermo Scientific Acros Supplier No. 435870010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 923590-95-8 | C8H8BrN·ClH | 234.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 923590-95-8
Molecular Formula C8H8BrN·ClH
Molecular Weight (g/mol) 234.52
InChI Key FQHLHVFOJBANKY-UHFFFAOYSA-N
Synonym 4-bromoisoindoline hydrochloride, 4-bromo-isoindoline hcl, 4-bromo-2,3-dihydro-1h-isoindole hydrochloride, 4-bromo-1h-isoindoline hydrochloride, 4-bromoisoindoline hcl, 1h-isoindole, 4-bromo-2,3-dihydro-, hydrochloride 1:1, pubchem14849, acmc-209y8q, 4-bromo-isoindoline hydrochloride
PubChem CID 44228644
IUPAC Name 4-bromo-2,3-dihydro-1H-isoindole;hydrochloride
SMILES C1C2=C(CN1)C(=CC=C2)Br.Cl

Tekniske data

CAS Min % 96.0
CAS Max % 100.0
Melting Point 269°C to 274°C
Color White to Yellow
Elemental Analysis 39.33 to 42.61 % C 5.73 to 6.21 % N
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 234.52
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromoisoindoline hydrochloride
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • 4-Bromoisoindoline hydrochloride
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sikkerhedssætning
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.