missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoisophthalic acid, 97%, Thermo Scientific™

Product Code. 10523511 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10523511 5 g Glass bottle 5g
10317470 25 g Glass bottle 25g
10762705 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10523511 Supplier Thermo Scientific Acros Supplier No. 151720050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6939-93-1
Molecular Formula C8H5BrO4
Molecular Weight (g/mol) 245.028
MDL Number MFCD00002404
InChI Key MSQIEZXCNYUWHN-UHFFFAOYSA-N
Synonym 4-bromoisophthalic acid, 1,3-benzenedicarboxylic acid, 4-bromo, 4-bromoisophthalicacid, 4-bromo-1,3-benzenedicarboxylic acid, 2-bromobenzene-1,5-dicarboxylic acid, pubchem12479, chemwish ic03530, ksc354c4d, msqiezxcnyuwhn-uhfffaoysa
PubChem CID 81361
IUPAC Name 4-bromobenzene-1,3-dicarboxylic acid
SMILES C1=CC(=C(C=C1C(=O)O)C(=O)O)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 297.0°C to 299.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Beilstein 09, 838
Formula Weight 245.03
Percent Purity 97%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-Bromoisophthalic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromoisophthalic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.