missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenol tert-Butyldimethylsilyl Ether, TRC

Product Code. 30332866
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30332866 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332866 Supplier Toronto Research Chemicals Supplier No. TRCB68644510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Bromophenol-tert-Butyldimethylsilyl Ether
CAS 67963-68-2
Molecular Formula C12 H19 Br O Si
Quantity 10 g
InChI Formula InChI=1S/C12H19BrOSi/c1-12(2,3)15(4,5)14-11-8-6-10(13)7-9-11/h6-9H,1-5H3
SMILES CC(C)(C)[Si](C)(C)Oc1ccc(Br)cc1
IUPAC Name (4-bromophenoxy)-tert-butyl-dimethylsilane
Molecular Weight (g/mol) 287.27
Formula Weight 286.0389
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.