missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylacetic Acid, TRC

Product Code. 30339658
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30339658 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339658 Supplier Toronto Research Chemicals Supplier No. TRCB68645025G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4-Bromophenyl)acetic Acid
CAS 1878-68-8
Molecular Formula C8 H7 Br O2
Quantity 25 g
Synonym Benzeneacetic acid, 4-bromo-, Acetic acid, (p-bromophenyl)- (7CI,8CI), 4-Bromobenzeneacetic acid, (4-Bromophenyl)acetic acid, (p-Bromophenyl)acetic acid, 2-(4-Bromophenyl)acetic acid, NSC 14358, 4-Bromophenylacetic acid
InChI Formula InChI=1S/C8H7BrO2/c9-7-3-1-6(2-4-7)5-8(10)11/h1-4H,5H2,(H,10,11)
SMILES OC(=O)Cc1ccc(Br)cc1
IUPAC Name 2-(4-bromophenyl)acetic acid
Molecular Weight (g/mol) 215.04
Formula Weight 213.9629
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.