missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromophenylethynyl)trimethylsilane, 98%

Product Code. 15421838
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15421838 25 g 25g
15401838 1 g 1g
15411838 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421838 Supplier Thermo Scientific Alfa Aesar Supplier No. H55534.14
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16116-78-2 | C11H13BrSi | 253.21 g/mol

It is used as the intermediate of Organic Light Emitting Diode (OLED).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as the intermediate of Organic Light Emitting Diode (OLED).

Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16116-78-2
Molecular Formula C11H13BrSi
Molecular Weight (g/mol) 253.21
MDL Number MFCD01863556
InChI Key RNMSGCJGNJYDNS-UHFFFAOYSA-N
Synonym 4-bromophenylethynyl trimethylsilane, 4-bromophenyl ethynyl trimethylsilane, 4-bromo-phenylethynyl-trimethyl-silane, 2-4-bromophenyl ethynyl trimethylsilane, acmc-1bvav, p-bromophenylethynyl trimethylsilane, 4-trimethylsilylethynyl bromo benzene, 4-bromo-phenylethynyl trimethyl silane
PubChem CID 4226980
IUPAC Name 2-(4-bromophenyl)ethynyl-trimethylsilane
SMILES C[Si](C)(C)C#CC1=CC=C(Br)C=C1

Specifications

Melting Point 61°C to 63°C
Color White to Yellow
Physical Form Powder
Quantity 25 g
Formula Weight 253.21
Percent Purity 98%
Chemical Name or Material (4-Bromophenylethynyl)trimethylsilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.