missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromophenylethynyl)trimethylsilane, 98%

Product Code. p-7052731
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15401838

Brand: Thermo Scientific Alfa Aesar H55534.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16116-78-2 | C11H13BrSi | 253.21 g/mol

It is used as the intermediate of Organic Light Emitting Diode (OLED).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as the intermediate of Organic Light Emitting Diode (OLED).

Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16116-78-2
Molecular Formula C11H13BrSi
Molecular Weight (g/mol) 253.21
MDL Number MFCD01863556
InChI Key RNMSGCJGNJYDNS-UHFFFAOYSA-N
Synonym 4-bromophenylethynyl trimethylsilane, 4-bromophenyl ethynyl trimethylsilane, 4-bromo-phenylethynyl-trimethyl-silane, 2-4-bromophenyl ethynyl trimethylsilane, acmc-1bvav, p-bromophenylethynyl trimethylsilane, 4-trimethylsilylethynyl bromo benzene, 4-bromo-phenylethynyl trimethyl silane
PubChem CID 4226980
IUPAC Name 2-(4-bromophenyl)ethynyl-trimethylsilane
SMILES C[Si](C)(C)C#CC1=CC=C(Br)C=C1

Specifications

Melting Point 61°C to 63°C
Color White to Yellow
Physical Form Powder
Quantity 1 g
Formula Weight 253.21
Percent Purity 98%
Chemical Name or Material (4-Bromophenylethynyl)trimethylsilane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.