missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophthalimide, 97+%

Product Code. 15481199
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15481199 5 g 5g
15471199 1 g 1g
15491199 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15481199 Supplier Thermo Scientific Alfa Aesar Supplier No. H64136.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6941-75-9 | C8H4BrNO2 | 226.029 g/mol

It is used as a intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a intermediate for pharmaceutical and organic synthesis.

Solubility
Soluble in N,N-DMF almost transparency.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6941-75-9
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.029
MDL Number MFCD00466049
InChI Key GNYICZVGHULCHE-UHFFFAOYSA-N
Synonym 4-bromophthalimide, 5-bromoisoindoline-1,3-dione, 5-bromo-isoindole-1,3-dione, 5-bromo-2,3-dihydro-1h-isoindole-1,3-dione, 4-bromo phthalimide, 5-bromo-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 5-bromo, 5-bromo-2h-isoindole-1,3-dione, 5-bromophthalimide, acmc-209o8b
PubChem CID 236018
IUPAC Name 5-bromoisoindole-1,3-dione
SMILES C1=CC2=C(C=C1Br)C(=O)NC2=O

Specifications

Melting Point 230°C to 235°C
Color White
Quantity 5 g
Beilstein 131235
Solubility Information Soluble in N,N-DMF almost transparency.
Formula Weight 226.03
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 4-Bromophthalimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.