missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromothien-3-yl)methanol, 97%, Thermo Scientific™

Product Code. 10603625
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10603625 250 mg Amber Glass Bottle 250mg
10767095 1 g Amber Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10603625

Brand: Thermo Scientific Maybridge CC54209CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 70260-05-8
Molecular Formula C5H5BrOS
Molecular Weight (g/mol) 193.06
MDL Number MFCD01859831
InChI Key MPKBPMYJCDNVEF-UHFFFAOYSA-N
Synonym 4-bromothiophen-3-yl methanol, 4-bromothien-3-yl methanol, 4-bromo-3-thiophenemethanol, 3-thiophenemethanol,4-bromo, 4-bromothiophen-3-yl ;methanol, 4-bromo-3-thienyl methan-1-ol, 3-bromo-4-hydroxymethyl thiophene
PubChem CID 818863
IUPAC Name (4-bromothiophen-3-yl)methanol
SMILES OCC1=CSC=C1Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 193.06
Percent Purity 97%
Chemical Name or Material (4-Bromothien-3-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.