missing translation for 'onlineSavingsMsg'
Learn More

4-Butoxybenzaldehyde, 99%, Thermo Scientific™

Product Code. 10124750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10124750 10 g Glass bottle 10g
10608824 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10124750 Supplier Thermo Scientific Acros Supplier No. 218370100
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 5736-88-9
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00003389
InChI Key XHWMNHADTZZHGI-UHFFFAOYSA-N
Synonym p-butoxybenzaldehyde, benzaldehyde, 4-butoxy, 4-n-butoxybenzaldehyde, benzaldehyde, p-butoxy, 4-butoxy-benzaldehyde, p-n-butoxybenzaldehyde, 4-n-butyloxybenzaldehyde, acmc-209lxn, p-n-butoxy benzaldehyde, 4-butoxybenzaldehyde
PubChem CID 79813
IUPAC Name 4-butoxybenzaldehyde
SMILES CCCCOC1=CC=C(C=C1)C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Red to Yellow
Boiling Point 148°C to 149°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)3OC6H4CHO
Refractive Index 1.5390 to 1.541
Quantity 10 g
Beilstein 08, II, 67
Formula Weight 178.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Butoxybenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.