missing translation for 'onlineSavingsMsg'
Learn More

4-Butoxybenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10601123
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
Product Code. Quantity unitSize
10601123 1 g 1g
10364962 10 g 10g
10640856 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 10601123

Brand: Thermo Scientific Maybridge TL00441DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1138-56-3
Molecular Formula C10H13ClO3S
Molecular Weight (g/mol) 248.721
MDL Number MFCD00052344
InChI Key HGKWMUBXVMFXNC-UHFFFAOYSA-N
Synonym 4-n-butoxy benzenesulfonyl chloride, 4-butoxybenzene-1-sulfonyl chloride, 4-n-butoxy benzenesulphonyl chloride, benzenesulfonyl chloride, 4-butoxy, 4-butoxy-benzenesulfonyl chloride, 4-n-butoxybenzenesulfonyl chloride, 4-butoxybenzenesulfonylchloride, pubchem5072, 4-n-butoxy benzenes, buttpark 9650-54
PubChem CID 517990
IUPAC Name 4-butoxybenzenesulfonyl chloride
SMILES CCCCOC1=CC=C(C=C1)S(=O)(=O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3(CH2)3OC6H4SO2Cl
Quantity 1 g
Formula Weight 248.73
Percent Purity 97%
Chemical Name or Material 4-Butoxybenzene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.