missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxy-9-acridanone (Technical Grade), TRC

Product Code. 30352606
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30352606 2.5 g 2.5g
30349006 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30352606 Supplier Toronto Research Chemicals Supplier No. TRCC1777782.5G
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24782-64-7
Molecular Formula C14H9NO3
Molecular Weight (g/mol) 239.23
Synonym 9,10-Dihydro-9-oxo-4-acridinecarboxylic Acid, 4-Carboxyacridone, 9(10H)-Acridone-4-carboxylic Acid, Acridone-4-carboxylic Acid, 9-oxo-4-acridancarboxylic Acid
IUPAC Name 9-oxo-10H-acridine-4-carboxylic acid
SMILES OC(=O)c1cccc2C(=O)c3ccccc3Nc12

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C14H9NO3/c16-13-8-4-1-2-7-11(8)15-12-9(13)5-3-6-10(12)14(17)18/h1-7H,(H,15,16)(H,17,18)
Formula Weight 239.0582
Chemical Name or Material 4-Carboxy-9-acridanone (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.