missing translation for 'onlineSavingsMsg'
Learn More

4-(Carboxymethyl)phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 11966061 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11966061 1 g 1g
11956061 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11966061 Supplier Thermo Scientific Acros Supplier No. 444670010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 797755-07-8
Molecular Formula C14H19BO4
Molecular Weight (g/mol) 262.11
MDL Number MFCD04038755
InChI Key FNLWHBHWDXCWHV-UHFFFAOYSA-N
Synonym 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetic acid, 4-carboxymethyl phenylboronic acid pinacol ester, phenylacetic acid-4-boronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetic acid, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylacetate, benzeneacetic acid, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-dihydroxyborane phenylacetic acid pinacol ester, 4-carboxymethyl phenylboronic acidpinacol ester
PubChem CID 3719002
IUPAC Name 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(CC(O)=O)C=C1

Specifications

Melting Point 166°C to 168°C
Color White to Yellow
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 262.11
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 4-(Carboxymethyl)phenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 4-(Carboxymethyl)phenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.