Learn More
Integra Biosciences™ 4-Channel Adapter
Product Code. 10563642
Change view
Click to view available options
For use with: Generic tip vacoboy
Product Code.
10563642 For use with: Generic tip vacoboy
1 options
Product Code. 10563642 Supplier Integra Biosciences™ Supplier No. 155524
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
VACUBOY hand operator set comprises of: VACUBOY hand operator, stand, rubber adapter for Pasteur pipettes, 1 channel stainless steel aspiration adapter 40mm, 8 channel stainless steel aspiration adapter 40mm, 1 channel plastic aspiration adapter for disposable tips, 1 channel plastic aspiration adapter for disposable tips with ejector, 8 channel plastic aspiration adapter for disposable tips with ejector. Can be used with VACUSAFE comfort vacuum system. Full range of accessories available. Enables liquid residues to be disposed of reliably, precisely and conveniently:
  • With a precisely dosed volume
  • Without loss of drops
  • Without risk of contamination
  • Without causing tiredness
  • Energy saving, the manual operator - the heart of the system - can be operated as a single or 8 channel aspirator
  • All parts in contact with fluids are autoclavable
Specifications
Includes Stainless Steel Tips
Length (Metric) Tip 40 mm
Type 4-channel Adapter