missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-1-methylpiperidine Hydrochloride, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-1-methylpiperidine Hydrochloride |
| CAS | 5382-23-0 |
| Molecular Formula | C6H13Cl2N |
| Quantity | 500 mg |
| Synonym | N-Methyl-4-chloropiperidine Hydrochloride, 1-Methyl-4-chloropiperidine Hydrochloride |
| InChI Formula | InChI=1S/C6H12ClN.ClH/c1-8-4-2-6(7)3-5-8;/h6H,2-5H2,1H3;1H |
| SMILES | Cl.CN1CCC(Cl)CC1 |
| IUPAC Name | 4-chloro-1-methylpiperidine;hydrochloride |
| Molecular Weight (g/mol) | 170.08 |
| Formula Weight | 169.0425 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction