missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(1H-pyrazol-3-yl)phenol, 98%, Thermo Scientific™

Product Code. 11347369
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11347369

Brand: Thermo Scientific Alfa Aesar H27482.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18704-67-1
Molecular Formula C9H7ClN2O
Molecular Weight (g/mol) 194.618
MDL Number MFCD00209566
InChI Key BAWPNSUPRVQKKN-UHFFFAOYSA-N
Synonym 4-chloro-2-1h-pyrazol-3-yl phenol, 3-5-chloro-2-hydroxyphenyl pyrazole, phenol,4-chloro-2-1h-pyrazol-3-yl, acmc-20amm6, dmglumyoloaxjy-uhfffaoysa-n, 2-1h-pyrazole-5-yl-4-chlorophenol, 4-chloro-2-1h-pyrazol-3-yl-phenol, phenol, 4-chloro-2-1h-pyrazol-3-yl, 4-chloro-2-2h-pyrazol-3-yl phenol, 4-chloro-6-1,2-dihydropyrazol-3-ylidene-1-cyclohexa-2,4-dienone
PubChem CID 6805776
IUPAC Name 4-chloro-6-(1,2-dihydropyrazol-3-ylidene)cyclohexa-2,4-dien-1-one
SMILES C1=CC(=O)C(=C2C=CNN2)C=C1Cl

Specifications

Melting Point 157°C to 158°C
Quantity 1 g
Formula Weight 194.62
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-(1H-pyrazol-3-yl)phenol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.