missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-isopropyl-5-methylphenol, 99%

Product Code. 11404987
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11404987 25 g 25g
11414987 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404987 Supplier Thermo Scientific Alfa Aesar Supplier No. B21906.14
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89-68-9 | C10H13ClO | 184.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-68-9
Molecular Formula C10H13ClO
Molecular Weight (g/mol) 184.66
MDL Number MFCD00002326
InChI Key KFZXVMNBUMVKLN-UHFFFAOYSA-N
Synonym chlorothymol, 6-chlorothymol, 4-chloro-2-isopropyl-5-methylphenol, chlorthymol, 4-chlorothymol, thymol, 6-chloro, phenol, 4-chloro-5-methyl-2-1-methylethyl, caswell no. 216, 4-chloro-6-isopropyl-3-methylphenol, unii-lj25ti0cvt
PubChem CID 6982
IUPAC Name 4-chloro-5-methyl-2-propan-2-ylphenol
SMILES CC(C)C1=CC(Cl)=C(C)C=C1O

Specifications

Melting Point 59°C to 61°C
Boiling Point 259°C to 263°C
Quantity 25 g
Beilstein 2084453
Sensitivity Light sensitive
Merck Index 14,2170
Formula Weight 184.67
Percent Purity 99%
Chemical Name or Material 4-Chloro-2-isopropyl-5-methylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.