missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylbenzoic Acid, TRC

Product Code. 30350676
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30350676 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350676 Supplier Toronto Research Chemicals Supplier No. TRCC42768525G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chloro-2-methylbenzoic Acid
CAS 7499-07-2
Molecular Formula C8H7ClO2
Quantity 25 g
Synonym 4-Chloro-o-toluic Acid, NSC 407521
InChI Formula InChI=1S/C8H7ClO2/c1-5-4-6(9)2-3-7(5)8(10)11/h2-4H,1H3,(H,10,11)
SMILES Cc1cc(Cl)ccc1C(=O)O
IUPAC Name 4-chloro-2-methylbenzoic acid
Molecular Weight (g/mol) 170.59
Formula Weight 170.0135
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.