missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylphenol, TRC

Product Code. 30354363
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30354363 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30354363

Brand: Toronto Research Chemicals TRCC37103010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chloro-2-methylphenol
CAS 1570-64-5
Molecular Formula C7 H7 Cl O
Quantity 10 g
Synonym o-Cresol, 4-chloro- (7CI,8CI), 4-Chloro-2-methylphenol, 4-Chloro-2-cresol, 4-Chloro-o-cresol, 4-Chlorocresol, 5-Chloro-2-hydroxytoluene, NSC 2851, PCOC, p-Chloro-o-cresol
InChI Formula InChI=1S/C7H7ClO/c1-5-4-6(8)2-3-7(5)9/h2-4,9H,1H3
SMILES Cc1cc(Cl)ccc1O
IUPAC Name 4-chloro-2-methylphenol
Molecular Weight (g/mol) 142.58
Formula Weight 142.0185
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.