missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylpyridine, 95%

Product Code. 15411139
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15411139 5 g 5g
15401139 1 g 1g
15421139 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15411139 Supplier Thermo Scientific Alfa Aesar Supplier No. H64093.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3678-63-5 | C6H6ClN | 127.571 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 3678-63-5
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.571
MDL Number MFCD00038833
InChI Key DAOZBJCTEPJGES-UHFFFAOYSA-N
Synonym 4-chloro-2-picoline, 2-methyl-4-chloropyridine, 4-chloro-2-methyl-pyridine, pyridine, 4-chloro-2-methyl, p-chloropicoline, 4-chloropicoline, 4-chloro-2-picolin, acmc-20amln, pubchem2071
PubChem CID 581392
IUPAC Name 4-chloro-2-methylpyridine
SMILES CC1=NC=CC(=C1)Cl

Spécification

Color Colorless to Yellow
Density 1.15 g/mL
Boiling Point 54°C to 56°C (15 mmHg)(literature)
Refractive Index 1.5205
Quantity 5 g
Beilstein 107196
Formula Weight 127.57
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-methylpyridine

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.