missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylquinoline, 97%

Product Code. 11467227
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467227 1 g 1g
11477227 5 g 5g
11487227 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467227 Supplier Thermo Scientific Alfa Aesar Supplier No. A15002.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4295-06-1 | C10H8ClN | 177.63 g/mol

4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4295-06-1
MDL Number MFCD00006757

Specifications

Melting Point ∼25°C
Density 0.881
Boiling Point 269°C to 270°C
Flash Point >110°C (230°F)
Molecular Formula C10H8ClN
Refractive Index 1.622
Quantity 1 g
Beilstein 3522
Synonym 4-chloroquinaldine, quinoline, 4-chloro-2-methyl, 4-chloro-2-methyl-quinoline, 2-methyl-4-chloroquinoline, quinaldine, 4-chloro, 4-chloro-quinaldine, acmc-20a2ui, 4-chloroquinaldine hcl, quinoline,4-chloro-2-methyl
Solubility Information Slightly soluble in water.
InChI Key HQAIROMRVBVWSK-UHFFFAOYSA-N
SMILES CC1=NC2=CC=CC=C2C(=C1)Cl
IUPAC Name 4-chloro-2-methylquinoline
Molecular Weight (g/mol) 177.63
PubChem CID 77973
Formula Weight 177.63
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methylquinoline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.