missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-methylquinoline, 97%
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Description
4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | ∼25°C |
| Density | 0.881 |
| Boiling Point | 269°C to 270°C |
| Flash Point | >110°C (230°F) |
| Molecular Formula | C9H6F3O3 |
| Refractive Index | 1.622 |
| Quantity | 5 g |
| Beilstein | 3522 |
| Synonym | 4-trifluoromethoxy phenylacetic acid, 2-4-trifluoromethoxy phenyl acetic acid, 4-trifluoromethoxy phenyl acetic acid, 4-trifluoromethoxy phenylaceticacid, 4-trifluoromethoxy-phenyl-acetic acid, 4-trifluoromethoxyphenylacetic acid, benzeneacetic acid, 4-trifluoromethoxy, maybridge3_006163 |
| Solubility Information | Slightly soluble in water. |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction