missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylthiopyrimidine, 97%

Product Code. 10552951 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10552951 50 g 50g
10347770 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10552951 Supplier Thermo Scientific Acros Supplier No. 163330500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 49844-90-8 | C5H5ClN2S | 160.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49844-90-8
Molecular Formula C5H5ClN2S
Molecular Weight (g/mol) 160.62
MDL Number MFCD00006083
InChI Key DFOHHQRGDOQMKG-UHFFFAOYSA-N
Synonym 4-chloro-2-methylthio pyrimidine, 4-chloro-2-methylthiopyrimidine, 4-chloro-2-methylsulfanyl pyrimidine, 4-chloro-2-methylsulfanyl-pyrimidine, pyrimidine, 4-chloro-2-methylthio, 4-chloro-2-methylmercapto pyrimidine, zlchem 575, pubchem15403, pubchem15764, acmc-1asbn
PubChem CID 97113
IUPAC Name 4-chloro-2-methylsulfanylpyrimidine
SMILES CSC1=NC=CC(Cl)=N1

Specifications

Melting Point -2.0°C
Density 1.3810g/mL
Boiling Point 139.0°C to 140.0°C (36.0 mmHg)
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6004
Quantity 50 g
Beilstein 23, 371
Specific Gravity 1.381
Formula Weight 160.63
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methylthiopyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloro-2-methylthiopyrimidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.