missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-2-nitroaniline, 99%

Product Code. 10167753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10167753 100 g Glass bottle 100g
10615772 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10167753 Supplier Thermo Scientific Acros Supplier No. 146521000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 89-63-4 | C6H5ClN2O2 | 172.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-63-4
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.57
MDL Number MFCD00007836
InChI Key PBGKNXWGYQPUJK-UHFFFAOYSA-N
Synonym benzenamine, 4-chloro-2-nitro, p-chloro-o-nitroaniline, pcon, 2-nitro-4-chloroaniline, red salt nbgl, devol red f, devol red salt f, red base ciba vi, red base irga vi, red salt ciba vi
PubChem CID 6979
IUPAC Name 4-chloro-2-nitroaniline
SMILES NC1=CC=C(Cl)C=C1[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 115°C to 119°C
Color Orange
Flash Point 191°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(NO2)NH2
Quantity 100 g
Beilstein 12, 729
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in methanol and ether
Formula Weight 172.57
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-chloro-2-nitroaniline
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Chloro-2-nitroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.