missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-phenylquinoline, 99%

Product Code. 10448900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10448900 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10448900 Supplier Thermo Scientific Acros Supplier No. 164870025
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4979-79-7 | C15H10ClN | 239.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4979-79-7
CAS Min % 98.5
CAS Max % 100.0
Melting Point 62°C to 64°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C15H10ClN
MDL Number MFCD00160641
Quantity 2.5 g
Synonym 4-chloro-2-phenyl-quinoline, 4-chloro-2-phenyquinoline, 2-phenyl-4-chloroquinoline, 4-chloro-2-phenyl-quinolin, 4-chloro-2-phenyl quinoline
InChI Key GLVDSTVYOFXBKT-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=NC3=CC=CC=C3C(=C2)Cl
IUPAC Name 4-chloro-2-phenylquinoline
Molecular Weight (g/mol) 239.71
PubChem CID 278376
Formula Weight 239.71
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Chloro-2-phenylquinoline, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.