missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-3-nitrobenzeneboronic acid pinacol ester, 98%

Product Code. 15460306
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460306 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15460306 Supplier Alfa Aesar™ Supplier No. H53022.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 913836-26-7
Molecular Formula C12H15BClNO4
Molecular Weight (g/mol) 283.52
MDL Number MFCD08235086
InChI Key VEIRGLFECZUCTI-UHFFFAOYSA-N
Synonym 2-4-chloro-3-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-chloro-3-nitrophenylboronic acid, pinacol ester, 4-chloro-3-nitrophenylboronic acid pinacol ester, 4-chloro-3-nitrobenzeneboronic acid, pinacol ester, 4-chloro-3-nitrobenzeneboronic acid,pinacol ester
PubChem CID 44119859
IUPAC Name 2-(4-chloro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC(=C(Cl)C=C1)[N+]([O-])=O

Specifications

Melting Point 56°C to 58°C
Quantity 250 mg
Formula Weight 283.52
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-nitrobenzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.