missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)pyridine hydrochloride, 97%

Product Code. 11338956
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11338956

Brand: Thermo Scientific Alfa Aesar L19556.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 732306-24-0 | C6H4Cl2F3N | 218.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 732306-24-0
Molecular Formula C6H4Cl2F3N
Molecular Weight (g/mol) 218.00
MDL Number MFCD04972770
InChI Key HCKXJUZITVCDND-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethyl pyridinehydrochloride, 4-chloro-3-trifluoromethyl pyridine, chloride, 4-chloro-3-trifluoromethyl pyridine hcl, 4-chloro-3-trifluoromethylpyridine hydrochloride, 3-chloro-4-trifluoromethyl pyridine hydrochlrorid, 4-chloranyl-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethylpyridine hydrochloride salt
PubChem CID 2782815
IUPAC Name 4-chloro-3-(trifluoromethyl)pyridine hydrochloride
SMILES Cl.FC(F)(F)C1=C(Cl)C=CN=C1

Specifications

Melting Point ∼135°C (sublimation)
Quantity 250 mg
Formula Weight 218.01
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)pyridine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.