missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-5-(2-thienyl)thieno[2,3-d]pyrimidine, 97%

Product Code. 15436457
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436457 1 g 1g
15448545 250 mg 250mg
15446457 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15436457 Supplier Thermo Scientific Alfa Aesar Supplier No. H33610.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 189681-04-7 | C10H5ClN2S2 | 252.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 189681-04-7
Molecular Formula C10H5ClN2S2
Molecular Weight (g/mol) 252.73
MDL Number MFCD00174019
InChI Key VNWBUFWVNCUUKY-UHFFFAOYSA-N
Synonym 4-chloro-5-2-thienyl thieno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-yl thieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine,4-chloro-5-2-thienyl, 4-chloro-5-2-thienyl thiopheno 2,3-d pyrimidine, 4-chloro-5-thiophen-2-ylthieno 2,3-d pyrimidine
PubChem CID 735732
IUPAC Name 4-chloro-5-thiophen-2-ylthieno[2,3-d]pyrimidine
SMILES ClC1=C2C(SC=C2C2=CC=CS2)=NC=N1

Specifications

Quantity 1 g
Formula Weight 252.75
Percent Purity 97%
Chemical Name or Material 4-Chloro-5-(2-thienyl)thieno[2,3-d]pyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.