missing translation for 'onlineSavingsMsg'
Learn More

4-chloro-5-nitro-o-phenylenediamine, 98%

Product Code. 10771444 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10771444 25 g Glass bottle 25g
10561622 1 g Glass bottle 1g
10610453 5 g Glass bottle 5g
11985261 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10771444 Supplier Thermo Scientific Acros Supplier No. 350660250
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 67073-39-6 | C6H6ClN3O2 | 187.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67073-39-6
Molecular Formula C6H6ClN3O2
Molecular Weight (g/mol) 187.58
MDL Number MFCD03427594
InChI Key LOQLMWFVXRZASN-UHFFFAOYSA-N
Synonym 4-chloro-5-nitro-o-phenylenediamine, 4-chloro-5-nitro-1,2-benzenediamine, 5-chloro-4-nitrobenzene-1,2-diamine, 4-chloro-5-nitro-1,2-phenylenediamine, 4-chloro-5-nitro-benzene-1,2-diamine, 1,2-benzenediamine, 4-chloro-5-nitro
PubChem CID 5702618
IUPAC Name 4-chloro-5-nitrobenzene-1,2-diamine
SMILES NC1=CC(Cl)=C(C=C1N)[N+]([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 189.0°C to 193.0°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 187.59
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-chloro-5-nitro-o-phenylenediamine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Chloro-5-nitro-o-phenylenediamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.