missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-6-iodoquinazoline, TRC

Product Code. 30349041
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349041 10 g 10g
30349043 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30349041 Supplier Toronto Research Chemicals Supplier No. TRCC36729510G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98556-31-1
Molecular Formula C8H4ClIN2
Molecular Weight (g/mol) 290.49
Synonym 6-Iodo-4-chloroquinazoline
IUPAC Name 4-chloro-6-iodoquinazoline
SMILES Clc1ncnc2ccc(I)cc12

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H4ClIN2/c9-8-6-3-5(10)1-2-7(6)11-4-12-8/h1-4H
Formula Weight 289.9108
Chemical Name or Material 4-Chloro-6-iodoquinazoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.