missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chloro-6-methoxymethyl-2-phenylpyrimidine

Product Code. 11355629
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11355629 250 mg 250mg
11365629 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355629 Supplier Alfa Aesar™ Supplier No. H50530.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Identificatori chimici

CAS 325685-59-4
Molecular Formula C12H11ClN2O
Molecular Weight (g/mol) 234.68
MDL Number MFCD00794769
InChI Key GSSSCZAOXFZADM-UHFFFAOYSA-N
Synonym 4-chloro-6-methoxymethyl-2-phenylpyrimidine, 6-chloro-2-phenyl-4-pyrimidinyl methyl methyl ether, acmc-20aml2, chlorophenylpyrimidinylmethylmethylether, 6-chloro-2-phenylpyrimidin-4-yl methoxymethane, pyrimidine,4-chloro-6-methoxymethyl-2-phenyl, 6-chloro-2-phenyl-4-pyrimidinyl methylmethyl ether
PubChem CID 2763927
IUPAC Name 4-chloro-6-(methoxymethyl)-2-phenylpyrimidine
SMILES COCC1=CC(Cl)=NC(=N1)C1=CC=CC=C1

Specifica

Quantity 250 mg
Formula Weight 234.69
Chemical Name or Material 4-Chloro-6-methoxymethyl-2-phenylpyrimidine
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.